missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Baffled Flask, with GL 45 thread

Product Code. 11726278 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Includes:
Membrane Cap and Pouring Ring
No. per Pack:
4/Pk.
Unit Size:
Each
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Includes No. per Pack Unit Size
11726278 250 mL Membrane Cap and Pouring Ring 4/Pk. Pack of 4
11736278 500 mL Membrane Cap and Pouring Ring 4/Pk. Pack of 4
15273436 1000 mL Membrane Cap and Pouring Ring 4/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11726278 Supplier DWK Life Sciences Supplier No. 212833655
  • ONLINE EXCLUSIVE PRICE
    124.00 €
    103.76 € / Pack of 4
    Save 20.24 €
    16% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Due to the baffles incorporated in the product, a solution to be shaken breaks on it and oxygen is introduced into the solution.

Oxygen intake is often the limiting factor for cell growth in the cultivation of microorganisms in Erlenmeyer flasks on a vibrating board or orbital shaker. The movement causes a liquid sickle to form when using DURAN™ Erlenmeyer flasks on a vibrator. The size of the sickle depends on the speed of the board and the vibration diameter. The greater the surface area of the contents, the greater the gas exchange area and therefore the potential oxygen intake. The speed and the associated oxygen intake can, however, only be increased to a limited extent. The new DURAN™ baffled flask with four baffles on the bottom disrupts the laminar flow and produces a turbulent flow. The surface area of the liquid and the gas exchange area are increased, thereby increasing the oxygen intake. Laboratory trials have demonstrated that the oxygen intake is doubled by the baffles compared to a standard DURAN™ Erlenmeyer flask. The Erlenmeyer flasks with baffles from the DWK Life Sciences can be geometrically reproduced due to completely automated and mechanical production. The wall thickness of the flasks was increased to achieve an excellent mechanical stability and to guarantee a long service life of the products. The special production process enables the manufacture of the product complete with thread in a two-stage process. The flasks can therefore be sealed with the tried-and-tested membrane screw cap from the DWK Life Sciences. This enables a reproducible gas exchange compared to other sealing mechanisms e.g. sealing with cotton wool.

  • Four baffled-bottom flasks cause a turbulent flow, increase the gas exchange surface and produce a higher oxygen transfer
  • At higher shaking speeds, the baffles increase aeration for better growth, especially with bacteria
  • The moulded bottom-baffles provide increased movement at lower speeds for a more gentle medium aeration of sensitive cultures
  • The flasks can therefore be sealed with the tried-and-tested membrane screw cap from the DWK Life Sciences which enables a reproducible gas exchange compared to other sealing mechanisms e.g. sealing with cotton wool
  • The flasks are manufactured in a fully automated process to provide totally consistent flask shapes leading to highly reproducible experimental conditions and results
  • The special production process enables the manufacture of the product complete with thread in a two-stage process
  • Available in range of three sizes for process scale up
  • The increased wall thickness of the flasks ensures excellent mechanical stability and a long service life.
  • Easy-to-read scale with graduation marks to estimate the content

Specifications

Diameter (Metric) Base 85 mm
Capacity (Metric) 250 mL
Material Borosilicate Glass (3.3)
For Use With (Application) Growth and cultivation of microorganisms; Media preparation and cultivation; Storage and transport of bacterial, yeast, plant and animal cells; Suitable for liquid cultures (e.g. pure cultures, enrichment cultures); Use on shaker equipment; GL 45 necks allowing easy use with : GL 45 membrane screw cap for use during autoclaving, 30 mm culture plugs or stoppers for culturing and GL 45 screw caps for media or reagent storage.
Includes Membrane Cap and Pouring Ring
No. per Pack 4/Pk.
Thread Size GL 45
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.