Learn More
DWK Life Sciences DURAN™ Blank for lid, flat flange, tall form, ground
Product Code. 15691594 Shop All DWK Life Sciences Products
Change view
Click to view available options
Description:
DN 100
DN 120
DN 150
DN 200
Unit Size:
Pack of 2
Pack of 3
Pack of 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
15691594 DN 100 Pack of 3
15291639 DN 150 Pack of 2
15201649 DN 200 Pack of 2
15281639 DN 120 Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15691594 Supplier DWK Life Sciences Supplier No. 210364609
  • ONLINE EXCLUSIVE PRICE
    262.50 €
    227.89 € / Pack of 3
    Save 34.61 €
    13% Off
Estimated Shipment: 28-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For customer-specific further processing.
DURAN™ blanks for flat flange covers can be further processed into customer-specific articles using further DURAN™ components (threaded tubes, standard ground joints, threaded sleeves, etc.). Due to their geometry and solid wall thickness, DURAN™ flat flange covers are pressure and vacuum resistant. For pressure resistance, please refer to our notes in the technical appendix of the laboratory glassware catalogue. For glass articles which deviate from the original form due to further processing, the permissible operating data must be determined in accordance with the technical rules and regulations.
  • High form, ground
  • Due to the geometry and the massive wall thickness, the blanks are suitable for devices with pressure and vacuum applications.
  • Glass articles for further processing
Specifications
Description DN 100
Material Borosilicate Glass (3.3)
For Use With (Application) For customer-specific further processing
No. per Pack 3/Pk.
Autoclavable Autoclavable