Learn More
Lenz Laborglasinstrumente™ Three-Neck Round Bottom Flasks, Side Necks Angled
Especially made for distillation, reactions and to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Three-Neck Round Bottom Flasks, Side Necks Angled are manufactured from durable DURAN™ borosilicate glass.
43.20 € - 408.10 €
Product Code | Brand | Capacity (Metric) | Socket Joint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Socket Joint | Price | Quantity & Availability | ||||
10478972
|
Lenz Laborglasinstrumente™
03313170 |
1000 mL | 14/23 NS Side Neck, 24/29 NS Center Neck |
87.34 €
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
||||
10742342
|
Lenz Laborglasinstrumente™
03314270 |
1000 mL | 19/26 NS Side Neck, 29/32 NS Center Neck |
69.75 €
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
||||
10129360
|
Lenz Laborglasinstrumente™
03314470 |
1000 mL | 29/32 NS Side Neck, 29/32 NS Center Neck |
121.00 €
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
10254381
|
Lenz Laborglasinstrumente™
03313237 |
100 mL | 19/26 NS Side Neck, 24/29 NS Center Neck |
58.19 €
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||||
10797511
|
Lenz Laborglasinstrumente™
03314437 |
100 mL | 29/32 NS Side Neck, 29/32 NS Center Neck |
94.18 €
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
10375211
|
Lenz Laborglasinstrumente™
03314237 |
100 mL | 19/26 NS Side Neck, 29/32 NS Center Neck |
55.24 €
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
10362701
|
Lenz Laborglasinstrumente™
03314149 |
250 mL | 14/23 NS Side Neck, 29/32 NS Center Neck |
43.20 €
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
10438402
|
Lenz Laborglasinstrumente™
03313149 |
250 mL | 14/23 NS Side Neck, 24/29 NS Center Neck |
65.24 €
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
10382701
|
Lenz Laborglasinstrumente™
03314249 |
250 mL | 19/26 NS Side Neck, 29/32 NS Center Neck |
52.07 €
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
10372701
|
Lenz Laborglasinstrumente™
03314449 |
250 mL | 29/32 NS Side Neck, 29/32 NS Center Neck |
54.96 €
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
11923401
|
Lenz Laborglasinstrumente™
03311122 |
25 mL | 14/23 NS Side Neck, 14/23 NS Center Neck |
54.22 €
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
10342751
|
Lenz Laborglasinstrumente™
03314158 |
500 mL | 14/23 NS Side Neck, 29/32 NS Center Neck |
45.95 €
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
10080211
|
Lenz Laborglasinstrumente™
03314482 |
5 L | 29/32 NS Side Neck, 29/32 NS Center Neck |
408.10 €
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
||||
10047570
|
Lenz Laborglasinstrumente™
03314258 |
500 mL | 19/26 NS Side Neck, 29/32 NS Center Neck |
59.63 €
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
10037570
|
Lenz Laborglasinstrumente™
03314458 |
500 mL | 29/32 NS Side Neck, 29/32 NS Center Neck |
58.30 €
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
Description
- Acc. to DIN 12394
- Two side necks at 20° angles
- With conical ground joints, featuring high taper accuracy and excellent tightness
- With inscription label
- Corrosion-free and resistant to thermal shock
Specifications
Borosilicate Glass, Duran Glass | |
Round |