Learn More
DWK Life Sciences DURAN™ Dimroth Condenser, with 2 standard ground joints, and 2 screw-on plastic hose connections
Product Code. 15630775 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint No:
NS 14/23
NS 24/29
NS 29/32
Length (Metric):
160 mm
250 mm
400 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) Unit Size
15630775 NS 14/23 160 mm Each
15640775 NS 24/29 250 mm Each
10040271 NS 29/32 250 mm Each
10080131 NS 29/32 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15630775 Supplier DWK Life Sciences Supplier No. 242546101
  • ONLINE EXCLUSIVE PRICE
    97.01 €
    87.72 € / Pack of 1
    Save 9.29 €
    9% Off
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Dimroth condenser with its internal cooling spiral has a large cooling surface and is often used as a reflux condenser.
Due to the internal cooling spiral, the Dimroth condenser is characterized by its large cooling surface (coolant flow in counter flow principle). The most well-known application of the Dimroth condenser is the reflux condenser. A reaction mixture is heated, the resulting vapours condense on the cooling spiral and are returned to the reaction mixture.
  • The Dimroth condenser comprises a coil condenser located within a tube
  • This condenser type has a larger heat exchange surface and thus a better cooling effect than the Liebig or Allihn condenser
  • Two removable plastic olives for inlet and outlet of the cooling medium
  • Tight closure due to standard ground joint
Specifications
For Use With (Application) Product and reflux condenser; Distillation; Condensers of length 160 mm (with standard ground joint 14/23) and 250 mm (with standard ground joint 24/29 and standard ground joint 29/32) are special sizes and not according to DIN
Length (Metric) 160 mm
Standard Taper Joint No NS 14/23
Material Borosilicate Glass (3.3), PBT, Polypropylene, Silicone
Type Non-DIN Size