missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Duran™ Extractors

Product Code. 11806194
Change view
Click to view available options
Capacity (Metric):
100 mL
200 mL
250 mL
1000 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11806194 250 mL Each
10587321 1000 mL Each
10615051 100 mL Each
10652361 200 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11806194 Supplier Lenz Laborglasinstrumente™ Supplier No. 05360049
  • ONLINE EXCLUSIVE PRICE
    336.00 €
    263.92 € / Pack of 1
    Save 72.08 €
    21% Off
View all promotions
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

For Use With (Equipment) Dimroth Condenser
Product Type Extractor
Capacity (Metric) 250 mL
Material Duran Glass
Includes Dimroth condenser
For Use With (Application) For the extraction from solids with solvents
Thread Size GL 14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.