Learn More
DWK Life Sciences DURAN™ Flat Flange, ground
Product Code. 15211639 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric) Base:
70 mm
106 mm
130 mm
155 mm
215 mm
Includes:
With groove
Without groove
Unit Size:
Pack of 2
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Base Includes Unit Size
15211639 106 mm With groove Pack of 2
11712909 70 mm With groove Pack of 2
11722909 130 mm With groove Pack of 2
11732909 130 mm Without groove Pack of 2
11860680 106 mm Without groove Pack of 2
15231639 215 mm With groove Pack of 2
15251639 155 mm Without groove Pack of 2
15261639 215 mm Without groove Pack of 2
15241639 70 mm Without groove Pack of 2
15221639 155 mm With groove Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15211639 Supplier DWK Life Sciences Supplier No. 210344607
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For processing as a reaction vessel.
DURAN™ flat flanges with and without grooves are suitable for further processing into pressure-resistant reaction vessels, for example. For pressure resistance, please refer to our notes in the technical appendix of the laboratory glassware catalogue. For glass articles which deviate from the original form due to further processing, the permissible operating data must be determined in accordance with the technical rules and regulations.
  • Ground
  • Component for further processing into reaction vessels
  • Due to the geometry and the massive wall thickness, the blanks are suitable for devices with pressure and vacuum applications.
  • Glass articles for further processing
Specifications
Diameter (Metric) Base 106 mm
Material Borosilicate Glass (3.3)
For Use With (Application) For processing as a reaction vessel
Includes With groove
No. per Pack 2/Pk.
Autoclavable Autoclavable