Learn More
DWK Life Sciences DURAN™ Folding Magnetic GL 45 Stirrer
Shop All DWK Life Sciences ProductsDescription
Self contained bottle stirring system ideal for simple mixing processes. Stirrer shaft can be adjusted to the optimal position in either a 500 or 1000 mL DURAN™ GL 45 bottles. Use with standard magnetic stirrers up to 500 rpm. Significantly improved mixing performance compared to standard magnetic stir bars. Two GL 14 port connector cap is compatible with the modular DURAN™ connection systems; tubing with 1.6 mm to 6.0 mm bores can be used to add or remove liquid or gas. Fully autoclavable. Parts in contact with media conform to FDA requirements. Supplied with or without bottle.
- The variable stirrer shaft can be adjusted to the optimal position in either 500 or 1000 ml DURAN™ GL 45 bottles
- Compatible with 1.6 to 6.0 mm outer diameter semi-rigid tubing for addition or removal of liquids or gas
- Use with standard magnetic stirrers up to 500 rpm and allows a clearly improved mixing
- Fully compatible with the DURAN™ connection systems
Specifications
Specifications
| Description | Folding magnetic stirrer for GL 45 stirred reactor, incl. Shaft |
| For Use With (Application) | Simple, low cost, bioreactor; Simple continuous flow stirred reactor; Mixing liquids, samples or reagents; Photo-bioreactor |
| Material | Polypropylene, PTFE, PEEK, Stainless Steel |
| Quantity | 1 Ea. |
| Thread Size (Metric) | GL 45 |
| Neck Finish | 20 mm |
| Autoclavable | Autoclavable |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.