missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Insert for Screw Cap GL 14, Silicone/PTFE (Screw Connection Cap GL 45 Accessory)

Product Code. 10523343 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric) Inner:
ID 1.6 mm
ID 3.0 mm
ID 3.2 mm
ID 6.0 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
10523343 ID 3.2 mm Each
10399652 ID 6.0 mm Each
10572763 ID 1.6 mm Each
10412153 ID 3.0 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10523343 Supplier DWK Life Sciences Supplier No. 1129817
Trusted Glassware Deals
Use promo code 28509
View all promotions
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Accessory for 2 or 3 port GL 45 connection cap system

Flexible modular GL 45 connection cap system that accepts four different rigid or semi-rigid tubing diameters (1.6 mm; 3.0 mm; 3.2 mm and 6.0 mm). The caps are available with 2 or 3 GL 14 threaded ports. The different sized inserts form a tight seal around the pass-through tubing. Sterile pressure equalisation is possible through use of 0.2 micron syringe filter. Unused ports can be sealed off with a red GL 14 screw cap.

  • Rigid or semi-rigid tubing of different diameters can be accepted by using a blue GL 14 open topped cap with one of the inserts
  • Available with 2, or 3 threaded outlets (ports)
  • Five different tubing diameters (3.2 mm, 6.0 mm, 8.0 mm, 10.0 mm and 12.0 mm) can be used by changing the inserts
  • By applying positive or negative pressure liquid can be pumped out of or into the bottle via the screw cap
  • Fully compatible with all the GL 45 threads and therefore highly versatile
  • Fully compatible with all the GLS 80 threads and therefore highly versatile

Suitable Accessory: 1129814

TRUSTED_SUSTAINABILITY

Specifications

Description Insert for screw cap GL 14, ID 3.2 mm (1/8 inch)
Diameter (Metric) Inner ID 3.2 mm
For Use With (Application) Sterile fluid transfer in and out of lab bottles; Gas transfer in and out of lab bottles; Fluid transfer between lab bottles; Supply of reagents to instruments; Bioreactor supply
Material Polypropylene, PTFE
Quantity 1 Ea.
Thread Size (Metric) GL 14
Diameter (Metric) 87 mm
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.