missing translation for 'onlineSavingsMsg'
Learn More

DWK LIFE SCIENCES™ DURAN™ Premium Bottle, Clear, with DIN 168-1 Thread, GL 45, Graduated

Product Code. 11786878 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11786878 100 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11786878 Supplier DWK LIFE SCIENCES™ Supplier No. 1127075
  • ONLINE EXCLUSIVE PRICE
    292.00 €
    280.88 € / Pack of 10
    Save 11.12 €
    3% Off
View all promotions
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The DURAN™ Premium Bottle, Clear, with DIN 168-1 Thread, GL 45, Graduated is moulded from USP/EP Type I borosilicate glass 3.3 with TÜV-certified 160 K thermal shock resistance and a PFA cap rated to 200°C.

  • Moulded from high-purity, clear USP/EP Type I borosilicate glass 3.3 with excellent thermal performance and high chemical resistance
  • TÜV-tested thermal shock resistance certified for a 160°C (160 K) differential
  • Complete with high-temperature PFA (fluoropolymer) screw cap and pouring ring rated for use up to 200°C
  • Standardised GL 45 thread and matching cap system for particularly tight sealing and simple, clean pouring
  • Double graduated scale for filling volume and a reversed scale for pouring (accurate to ± 5%)
  • Marking field and Retrace Code for manufacturing lot traceability printed in permanent white enamel
  • Colourant-free product; PTFE-coated cap liner ensures reliable sealing
  • Robust ISO 4796-1 design for a long working life

Specifications

Capacity (Metric) 100 mL
Color Clear
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage and transport of media and reagents; Shipping
Material Borosilicate Glass (3.3)
Sterility Non-sterile
Thread Size (Metric) GL 45
Closure Included Yes
Graduated Yes
Mouth Narrow
Height (Metric) 105 mm
Chemical Label Unlabeled
Diameter (Metric) 56 mm
Autoclavable Autoclavable
Capacity (English) 3 oz.
Shape Square
Disposable Reusable
Product Type Reagent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.