Learn More
Lenz Laborglasinstrumente™ Duran Glass Bent Gas Inlet Tubes
Product Code. 10250881
Change view
Click to view available options
Length (Metric):
250mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
10250881 250mm Each
1 options
Product Code. 10250881 Supplier Lenz Laborglasinstrumente™ Supplier No. 05083001
Edge
Specifications
Includes NS cone and thin capillary
Length (Metric) 250mm
Type Gas Inlet Tube
Material Duran Glass
Quantity 1Each