Learn More
DWK Life Sciences DURAN™ GLS 80™ Pouring Ring, Blue PP
Product Code. 10611724 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GLS 80
Unit Size:
Pack of 10
Product Code. Thread Size (Metric) Unit Size
10611724 GLS 80 Pack of 10
1 options
Product Code. 10611724 Supplier DWK Life Sciences Supplier No. 1160166
  • ONLINE EXCLUSIVE PRICE
    29.62 €
    22.30 € / Pack of 10
    Save 7.32 €
    24% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Blue PP GLS 80™ wide-mouth screw cap (plug seal) and drip free, PP pouring ring for GLS 80™ bottles
Permits opening and closing of the DURAN™ GLS 80™ bottle with only a three-quarter turn. A matching PP pouring ring is also available, permitting clean, drip-free use.
  • Triple thread permits user friendly opening and closing with only a three-quarter turn
  • Cap offer a linerless, integral lip seal
  • Pouring ring for drip-free pouring and clean, safe working
Specifications
Description Pouring ring
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping
Material Polypropylene
Quantity 10/Pk.
Thread Size (Metric) GLS 80
Neck Finish 16 mm
Diameter (Metric) 87 mm
Autoclavable Autoclavable