Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, flat, diamond-ground
Product Code. 15991521 Shop All DWK Life Sciences Products
Change view
Click to view available options
Size:
NS 10/19
NS 12/21
NS 14/23
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 45/40
NS 7/16
Unit Size:
Each
Pack of 10
Pack of 100
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15991521 NS 19/26 Pack of 100
10079940 NS 10/19 Pack of 100
10460873 NS 12/21 Pack of 100
15814558 NS 24/29 Pack of 100
15901531 NS 29/32 Pack of 100
10768631 NS 34/35 Each
15931531 NS 45/40 Pack of 10
15971521 NS 14/23 Pack of 100
15931521 NS 7/16 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15991521 Supplier DWK Life Sciences Supplier No. 286126206
  • ONLINE EXCLUSIVE PRICE
    243.00 €
    208.56 € / Pack of 100
    Save 34.44 €
    14% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Hollow glass stopper for sealing.
DURAN™ hexagonal hollow plugs are used for sealing laboratory equipment such as volumetric flasks, mixing cylinders, ground joint flasks, etc. DURAN™ hexagonal hollow plugs are available in 2 standard versions - pointed - flat - Sizes analogous to DURAN™ conical ground joints. Cut quality: DG = Diamond Ground
  • Flat
  • Diamond-ground
  • hollow glass stopper
Specifications
For Use With (Application) Closure of vessels with standard ground joints
Quantity 100/Pk.
Size NS 19/26
Autoclavable Autoclavable
Material Borosilicate Glass