missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Membrane Venting GL Screw Cap, Blue PP, Welded-in ePTFE Membrane for Pressure Equalisation During Autoclaving
Shop All DWK Life Sciences ProductsDescription
- Pressure equalisation during steam sterilisation, the 0.2μm ePTFE membrane permits automatic sterile gas exchange in two directions
- Microporous ePTFE membrane blocks ingress of liquids or solids, keeping bottle contents sterile throughout the autoclave cycle
- Increased gas exchange rate due to thin 0.2μm ePTFE membrane combined with larger ventilation openings
- PTFE membrane and polypropylene mesh support are ultrasonically welded in place, forming a durable seal
- Reusable and cost effective — replacement recommended after 3 to 5 autoclave cycles or if membrane becomes damaged or contaminated
- ePTFE membrane conforms to USP 121°C Class VI; supported by polypropylene mesh
- Compatible with GL 25, GL 32, and GL 45 DIN thread laboratory bottles; ideal for storage or transport of gas-generating media
Specifications
Specifications
| Color | Blue |
| For Use With (Application) | Pressure equalisation during steam sterilisation/autoclaving of media or buffers; Sterile gas exchange for aerobic microbial, shaker and suspension cell cultures; Autoclaving of media and buffers; Short-term storage of sterilised media; Storage of gas generating materials |
| Material | Polypropylene, ePTFE |
| Quantity | 5/Pk. |
| Sterility | Non-sterile |
| Thread Size (Metric) | GL 25 |
| Diameter (Metric) | 33 mm |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Product Type | Bottle Cap |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?