missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Microbiology Bottle, Narrow Neck, Unground
Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
20,000 mL
2000 mL
25 mL
250 mL
50 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
Description
The culture bottles are moulded from DURAN™ borosilicate 3.3 Type 1 glass, and are suitable for the preparation of media or culturing of microorganisms. The bottles comply with ISO 4796-2 and feature a narrow conical unground neck for use with rubber stoppers. Suitable for autoclaving at 121 or 134°C or dry heat sterilization.
- Narrow neck for rubber stoppers
- Retrace code
- Autoclavable at 121°C or 134°C
- Supplied without stopper
Specifications
Specifications
| Capacity (Metric) | 5000 mL |
| For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Culturing vessel |
| Includes | With print |
| Material | Borosilicate Glass (3.3) |
| Quantity | 1 Ea. |
| Diameter (Metric) | 182 mm |
| Autoclavable | Autoclavable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction