missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Microbiology Bottle Unground Wide Neck

Product Code. 15251659 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
20,000 mL
2000 mL
250 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15251659 250 mL Pack of 10
10261701 5000 mL Each
10211801 20,000 mL Each
11702919 500 mL Pack of 10
11800690 1000 mL Pack of 10
10363351 2000 mL Pack of 10
10313451 10,000 mL Each
15241659 100 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15251659 Supplier DWK Life Sciences Supplier No. 211813601
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Culture bottles with a wide neck for use with rubber stoppers

The culture bottles are moulded from DURAN™ borosilicate 3.3 Type 1 glass, and are suitable for the preparation of media or culturing of microorganisms. The bottles comply with ISO 4796-2 and feature a wide conical unground neck for use with rubber stoppers. Suitable for autoclaving at 121 or 134°C or dry heat sterilization.

  • Wide neck for rubber stoppers
  • Wide neck for easier filling and cleaning
  • Retrace code
  • Fired-on, highly durable white print
  • Autoclavable at 121° C or 134°C
  • Supplied without stopper
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 250 mL
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Culturing vessel
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Diameter (Metric) 70 mm
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.