Learn More
DWK Life Sciences DURAN™ Original Laboratory Bottle, Amber, with DIN 168-1 Thread, USP / EP (3.2.1) Light Transmission, Graduated
Shop All DWK Life Sciences ProductsDescription
DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The bottles are moulded from high-purity, amber, USP/EP Type I DURAN™ borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The addition of a fused, external amber layer ensures that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards and lasts the lifetime of the bottle. The amber bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ Original amber bottles are supplied with or without iconic blue polypropylene cap and drip-free pouring ring for use up to 140°C. All components are fully autoclavable at 121 or 134°C.
- Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
- UV protection up to approx. 500 nm due to external colouration
- The only external application of the colour ensures that internal surfaces have standard DURAN™ glass properties
- Very durable and chemically resistant amber colour
- Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic print
- Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required
Specifications
Specifications
| Capacity (Metric) | 5000 mL |
| Color | Amber |
| For Use With (Application) | Preparation of light-sensitive materials; Storage of light-sensitive materials; Transport and shipping of light-sensitive materials; Autoclaving of media and reagents; Culturing vessel |
| Includes | Blue Polypropylene Screw Cap and Pouring Ring |
| Material | Borosilicate Glass (3.3) |
| No. per Pack | 1 Ea. |
| Quantity | 1 Ea. |
| Sterility | Non-sterile |
| Thread Size (Metric) | GL 45 |
| Closure Color | Blue |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.