missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated

Product Code. p-7189449 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
150 mL
250 mL
500 mL
750 mL
1000 mL
2 L
3500 mL
5000 mL
10 L
15 L
20 L
25 L
Unit Size:
Each
Pack of 10
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11752282 25 mL Pack of 10
11782282 50 mL Pack of 10
11772282 100 mL Pack of 10
11762282 250 mL Pack of 10
11752272 500 mL Pack of 10
12347178 1000 mL Pack of 10
10724052 3500 mL Each
11699888 150 mL Pack of 10
10119810 10 L Each
11619898 750 mL Pack of 10
10038050 5000 mL Each
13407119 10 mL Pack of 10
10047990 20 L Each
15233719 15 L Each
15806881 25 L Each
11792282 2 L Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Product Code. 11752282 Supplier DWK Life Sciences Supplier No. 218011453
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The gold standard for multifunctional glass laboratory bottles.

  • Complies with the standards in ISO 4796-1:2016 Laboratory glassware - Bottles - Part 1: Screw-neck bottles (does not apply to 10 mL bottle)
  • Manufactured from DURAN™ borosilicate glass 3.3
  • DURAN™ is a neutral glass, classified as Glass Type I according to USP <660>, EP (3.2.1), and JP <7.01>
  • Meets ASTM E438, Type I, Class A requirements
  • Standardized GL-thread and matching cap system for especially tight sealing and simple, clean pouring
  • Retrace Code and certificate available for manufacturing lot traceability (downloadable via DWK certificate system)
  • Fired-on, highly durable white print
  • Suitable for long-term storage
  • Easy-to-read scale with clear graduation marks for estimating contents

Specifications

Capacity (Metric) 25 mL
Color Clear
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir
Includes Blue Polypropylene Screw Cap only
Material Borosilicate Glass (3.3)
No. per Pack 10/Pk.
Quantity 10/Pk.
Sterility Non-sterile
Thread Size (Metric) GL 25
Closure Color Blue
Closure Included Yes
Graduated Yes
Mouth Narrow, Threaded
Fit for Purpose Certified Quality
Height (Metric) 74 mm
Closure Liner Linerless
Closure Material Polypropylene
Closure Size GL 25
Diameter (Metric) 36 mm
Autoclavable Autoclavable
Shape Round
Disposable Reusable
Product Type Laboratory Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.