missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Original Laboratory GL Bottle Screw Cap, Linerless Lip Seal, PP

Product Code. p-7189113 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GL 25
GL 32
GL 45
Color:
Blue
Gray
Green
Yellow
Unit Size:
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) Color unitSize
11725434 GL 45 Yellow Pack of 10
11722322 GL 25 Blue Pack of 10
11732322 GL 32 Blue Pack of 10
11762332 GL 45 Blue Pack of 10
11735434 GL 45 Green Pack of 10
11745434 GL 45 Gray Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11725434 Supplier DWK Life Sciences Supplier No. 293382802
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Screw cap with reliable plug seal.

The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.

  • Cap with integral lip seal provides effective sealing for short term storage
  • The colour helps prevent interchanging of cap mix-ups and reduces the risk of cross contamination
  • Fully compatible with all the GL 45 threads and therefore highly versatile
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description Screw cap
For Use With (Application) Autoclaving media and reagents; Short term sample and media storage; color coding and identification
Material Polypropylene
Quantity 10/Pk.
Thread Size (Metric) GL 45
Diameter (Metric) 54 mm
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.