missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ DURAN™ protect+ Graduated Amber Laboratory Bottle with DIN 168-1 Thread

Product Code. 17976792 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1 L
2 L
5 L
Thread Size (Metric):
GL 25
GL 32
GL 45
Unit Size:
Each
Pack of 10
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Thread Size (Metric) Unit Size
17976792 5 L GL 45 Each
17976782 25 mL GL 25 Pack of 10
17946772 50 mL GL 32 Pack of 10
17996782 100 mL GL 45 Pack of 10
17906792 250 mL GL 45 Pack of 10
17936792 500 mL GL 45 Pack of 10
17956792 1 L GL 45 Pack of 10
17966792 2 L GL 45 Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 17976792 Supplier DWK Life Sciences™ Supplier No. 218067382
  • ONLINE EXCLUSIVE PRICE
    649.00 €
    522.48 € / Pack of 1
    Save 126.52 €
    19% Off
View all promotions
Estimated Shipment: 25-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Multi-functional ambered glass laboratory bottles with UV protection and a protective safety coating.

DURAN™ GL ISO bottles have become ideal multi-functional laboratory bottles since their introduction in 1972. The protect+ safety-coated version of the bottles combines the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 on the inside and a protective safety plastic layer on the outside. The bottles are molded from high-purity, clear glass or externally ambered clear glass that is then coated with a translucent polyamide PA 12 coating to reduce the hazards of broken glass and liquid splashes in the event of breakage.

A combination of high-performance materials and robust design provides a cost-effective usable life. The bottles feature an easy-to-read graduated volume scale. There is a Retrace Code printed on each bottle for manufacturing lot traceability. The ambered DURAN™ glass bottle versions ensure that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards. All the protect+ coated bottles are supplied with push-on protective covers to protect the inside of the bottles from contamination during distribution. Matching screw closures and pouring rings can be ordered separately. Available in a wide range of nominal bottle capacities from 25 mL to 5 L, and neck sizes GL 25 to GLS 80™ wide mouth.

  • Manufactured from ambered borosilicate glass 3.3 and polyamide
  • Standardized GL-thread compatible with matching cap system for particularly tight sealing and simple, clean pouring out
  • The protect+ safety coating provides scratch, leak* and splinter* protection (*only applies to bottles 2000 mL or less)
  • UV protection USP (Spectral Transmission) and EP (3.2.1) compliant
  • Less than 10% spectral transmission between 290 nm and 450 nm
  • The only external application of the amber color and safety coating ensures that internal surfaces have standard DURAN™ glass properties
  • Supplied as safety coated glass bottle only (screw caps and pouring rings must be ordered separately)
  • Easy-to-read scale with graduation marks to estimate the content
  • The 25 mL has a specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required
  • Safe storage and shipping of bulk, high-potency active pharmaceutical ingredients (HPAPIs) powders
  • Coating process does not use any volatile organic compounds (VOCs) or isocyanates

Specifications

Capacity (Metric) 5 L
Color Amber
For Use With (Application) Preparation of light-sensitive media and reagents, safe storage and shipping of bulk, light-sensitive, high-potency active pharmaceutical ingredients (HPAPIs) powders, sample or harvest collection container, sulturing vessel, storage of light-sensitive media and reagents, transport and shipping
Material Borosilicate Glass (3.3)
Quantity 1 Bottle
Sterility Non-sterile
Weight (Metric) 2.05 kg
Thread Size (Metric) GL 45
Closure Included No
Graduated Yes
Mouth Wide
Height (Metric) 330 mm
Closure Size 45
Diameter (Metric) 182 mm
Autoclavable Not Autoclavable
Capacity (English) 169.1 oz.
Shape Round
Certifications/Compliance CoC
Product Type Laboratory Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.