missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ DURAN™ protect+ Graduated Amber Laboratory Bottle with Wide Mouth

Product Code. 17936762 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1 L
2 L
3.5 L
5 L
Unit Size:
Each
Pack of 10
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17936762 3.5 L Each
17976752 250 mL Pack of 10
17986752 500 mL Pack of 10
17916762 1 L Pack of 10
17946762 2 L Pack of 6
17956762 5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17936762 Supplier DWK Life Sciences™ Supplier No. 218666983
  • ONLINE EXCLUSIVE PRICE
    427.00 €
    415.87 € / Pack of 1
    Save 11.13 €
    2% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Multi-functional glass wide-mouth laboratory bottles with protective safety coating.

The protect+ safety-coated version of the bottles combines the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 on the inside and a protective safety plastic layer on the outside. The bottles are molded from high-purity, clear glass or externally ambered clear glass that is then coated with a translucent polyamide PA 12 coating to reduce the hazards of broken glass and liquid splashes in the event of breakage.

A combination of high-performance materials and robust design provides a cost-effective usable life. The bottles feature an easy-to-read graduated volume scale. There is a Retrace Code printed on each bottle for manufacturing lot traceability. The ambered DURAN™ glass bottle versions ensure that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards. All the protect+ coated bottles are supplied with push-on protective covers to protect the inside of the bottles from contamination during distribution. Matching screw closures and pouring rings can be ordered separately. Available in a wide range of nominal bottle capacities from 25 mL to 5 L, and neck sizes GL 25 to GLS 80™ wide mouth.

  • Manufactured from ambered borosilicate glass 3.3 and polyamide
  • Compatible with matching cap system for particularly tight sealing and simple, clean pouring out
  • protect+ safety coating provides scratch, leak* and splinter* protection (*only applies to bottles 2000 mL or less)
  • UV protection USP (Spectral Transmission) and EP (3.2.1) compliant
  • Less than 10% spectral transmission between 290 nm and 450 nm
  • The only external application of the amber colour and safety coating ensures that internal surfaces have standard DURAN™ glass properties
  • Supplied as safety coated glass bottle only (screw caps and pouring rings must be ordered separately)
  • Easy-to-read scale with graduation marks to estimate the content
  • Safe storage and shipping of bulk, high-potency active pharmaceutical ingredients (HPAPIs) powders
  • Coating process does not use any volatile organic compounds (VOCs) or isocyanates

Specifications

Capacity (Metric) 3.5 L
Color Amber
For Use With (Application) Preparation of light-sensitive media and reagents, safe storage and shipping of bulk, light-sensitive, high-potency active pharmaceutical ingredients (HPAPIs) powders, sample or harvest collection container, sulturing vessel, storage of light-sensitive media and reagents, transport and shipping
Material Borosilicate Glass (3.3)
Quantity 1 Bottle
Sterility Non-sterile
Weight (Metric) 1.377 kg
Thread Size (Metric) GLS 80
Closure Included No
Graduated Yes
Mouth Wide
Height (Metric) 271 mm
Closure Size 80
Diameter (Metric) 160 mm
Autoclavable Not Autoclavable
Capacity (English) 118.3 oz.
Shape Round
Certifications/Compliance CoC
Product Type Laboratory Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.