missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ DURAN™ protect+ Graduated Laboratory Bottles with DIN 168-1 Thread
GREENER_CHOICE

Product Code. 17996792 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1 L
2 L
3.5 L
5 L
Thread Size (Metric):
GL 25
GL 32
GL 45
Unit Size:
Each
Pack of 10
Pack of 6
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Thread Size (Metric) Unit Size
17996792 5 L GL 45 Each
17906772 25 mL GL 25 Pack of 10
17926772 50 mL GL 32 Pack of 10
17956772 100 mL GL 45 Pack of 10
17986782 250 mL GL 45 Pack of 10
17916792 500 mL GL 45 Pack of 10
17946792 1 L GL 45 Pack of 10
17926792 2 L GL 45 Pack of 6
17986792 3.5 L GL 45 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 17996792 Supplier DWK Life Sciences™ Supplier No. 218057381
  • ONLINE EXCLUSIVE PRICE
    434.76 €
    397.03 € / Pack of 1
    Save 37.73 €
    8% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Multi-functional clear glass laboratory bottles with protective safety coating.

DURAN™ GL ISO bottles have become ideal multi-functional laboratory bottles since their introduction in 1972. The protect+ safety-coated version of the bottles combines the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 on the inside and a protective safety plastic layer on the outside. The bottles are molded from high-purity, clear glass or externally ambered clear glass that is then coated with a translucent polyamide PA 12 coating to reduce the hazards of broken glass and liquid splashes in the event of breakage.

A combination of high-performance materials and robust design provides a cost-effective usable life. The bottles feature an easy-to-read graduated volume scale. There is a Retrace Code printed on each bottle for manufacturing lot traceability. The ambered DURAN™ glass bottle versions ensure that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards. All the protect+ coated bottles are supplied with push-on protective covers to protect the inside of the bottles from contamination during distribution. Matching screw closures and pouring rings can be ordered separately. Available in a wide range of nominal bottle capacities from 25 mL to 5 L, and neck sizes GL 25 to GLS 80™ wide mouth.

  • Manufactured from polyamide-coated DURAN™ borosilicate glass 3.3
  • DURAN™ is a neutral glass, classified as a Glass Type I according to USP, EP (3.2.1) and JP, and meets ASTM E438, Type I, Class A requirements
  • Reduce the hazards of broken glass and liquid splashes in the event of breakage
  • External polymer coating provides scratch, leak* and splinter* protection (*only applies to bottles 2000 mL or less)
  • Polyamide 12 safety coating has a high chemical resistance
  • Protective coating provides an enhanced safer grip
  • Coating is Cleanroom certified Fraunhofer IPA TESTED DEVICE™
  • Certified FDA and EU food contact safe coating material
  • Safe storage and shipping of bulk, high-potency active pharmaceutical ingredients (HPAPIs) powders
  • Supplied with push-on dust cover
  • Coating process does not use any volatile organic compounds (VOCs) or isocyanates
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 5 L
Color Clear
For Use With (Application) Preparation of media and reagents, safe storage and shipping of bulk, high-potency active pharmaceutical ingredients (HPAPIs), sample or harvest collection container, culturing vessel, storage of media and reagents, transport and shipping, Liquid handling reservoir
Material Borosilicate Glass (3.3)
Quantity 1 Bottle
Sterility Non-sterile
Weight (Metric) 2.05 kg
Thread Size (Metric) GL 45
Closure Included No
Graduated Yes
Mouth Wide
Height (Metric) 330 mm
Closure Size 45
Diameter (Metric) 182 mm
Autoclavable Not Autoclavable
Capacity (English) 169.1 oz.
Shape Round
Certifications/Compliance CoC
Product Type Laboratory Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.