Learn More
DWK Life Sciences DURAN™ PURE Premium Screw Cap
BIOPHARMA
Product Code. p-7189419 Shop All DWK Life Sciences Products
Click to view available options
Thread Size (Metric):
GL 25
GL 45
Unit Size:
Pack of 5
Product Code. 11679808
Brand: DWK Life Sciences 891088679
Only null left
Add to basket
Edge
Add to basket
High-quality closures developed for the needs of the pharmaceutical and biotech industry
High-quality plastic screw caps with GL 25/GL 45 thread sizes, developed for the needs of the pharmaceutical industry. DURAN™ PURE Premium Screw Caps are manufactured in clean rooms and made from PFA with highly inert PTFE coated silicone seal. Designed for use with DURAN™ PURE GL 25/ GL 45 Bottles. The screw caps are compatible with autoclaving and other common sterilization methods. DWK Life Sciences provides CoAs with each delivery, assures 100% traceability and change control management. DURAN™ PURE Premium Screw Caps are available with the documentation required by regulatory authorities and meet the requirements of the international pharmacopoeias.
  • High cleanliness due to manufacturing in clean rooms
  • Single batch delivery
  • Certificate of Analysis (CoA)
  • Visual and dimensional inspection
  • Change control management and information of customer in case of changes
  • 100% traceability of all manufacturing steps
  • Extractables report available
  • Drug Master File
  • USP <88> Class VI Pharmaceutical grade materials
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
For Use With (Application) Manufacturing of pharmaceutical drugs and biotech products; Storage and transport of APIs, bulk drug intermediates, ingredients and media
Material PFA, PTFE, Silicone
Quantity 5/Pk.
Thread Size (Metric) GL 45
Diameter (Metric) 51 mm
Autoclavable Autoclavable