missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Reagent Bottle, Narrow Neck, Amber, USP /EP (3.2.1) Light Transmission

Product Code. 11786248 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
20,000 mL
2000 mL
25 mL
250 mL
50 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11786248 2000 mL Pack of 10
11766858 1000 mL Pack of 10
11746858 250 mL Pack of 10
11736858 100 mL Pack of 10
11756858 500 mL Pack of 10
11726858 50 mL Pack of 10
10078000 5000 mL Each
10672252 10,000 mL Each
11786268 25 mL Pack of 10
11689858 20,000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 11786248 Supplier DWK Life Sciences Supplier No. 211686309
  • ONLINE EXCLUSIVE PRICE
    1730.00 €
    1697.22 € / Pack of 10
    Save 32.78 €
    1% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Classic narrow-neck, glass-stoppered reagent bottles in ambered glass.

The classical stoppered reagent bottles are moulded from amber DURAN™ borosilicate 3.3 Type 1 glass, and are suitable for the storage of liquid and solid chemicals and reagents. The bottles feature a narrow conical ground joint neck and comply with ISO 4796-2. The narrow neck is suitable for liquids and the angle of the shoulders ensures that the contents can be smoothly poured. The glass stopper provides an excellent seal that protects the contents from evaporation, oxidation, and contamination. The bottles have a Retrace Code for manufacturing lot traceability and a marking field in permanent white enamel. The amber glass ensures that light sensitive contents are protected from the effects of photo-degradation. The protection against ultra-violet light meets international USP/EP standards and lasts the lifetime of the bottle.

  • USP/EP UV protection up to approx. 500 nm due to external colouration
  • The amber colour is only applied externally ensuring that internal surfaces have standard DURAN™ clear Type 1 glass properties
  • Very durable and chemically resistant amber colour
  • All glass components and therefore suitable for storage of aggressive materials
  • Tight closure due to standard ground joint
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic print

Specifications

Capacity (Metric) 2000 mL
Description Neck with standard ground joint and glass flat-headed stopper
For Use With (Application) Storage of air sensitive reagents, and chemicals; Storage of hygroscopic liquids, powders and granules; Storage of air sensitive reagents and chemicals; Storage of hygroscopic liquids and pharmaceuticals
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Diameter (Metric) 134 mm
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.