missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Reagent Bottle, Narrow Neck, Clear

Product Code. 11722262 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10 mL
10,000 mL
100 mL
1000 mL
20,000 mL
2000 mL
25 mL
250 mL
50 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11722262 250 mL Pack of 10
10528871 10,000 mL Each
11716858 1000 mL Pack of 10
11706858 500 mL Pack of 10
10363781 5000 mL Each
11756928 25 mL Pack of 10
11746948 2000 mL Pack of 10
15231659 10 mL Pack of 10
11786848 50 mL Pack of 10
11361465 100 mL Pack of 10
10645141 20,000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 11722262 Supplier DWK Life Sciences Supplier No. 211653609
  • ONLINE EXCLUSIVE PRICE
    432.00 €
    385.25 € / Pack of 10
    Save 46.75 €
    10% Off
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Classic narrow-neck, glass-stoppered reagent bottles.

The classical stoppered reagent bottles are moulded from DURAN™ borosilicate 3.3 Type 1 glass, and are suitable for the storage of liquid and solid chemicals and reagents. The bottles feature a narrow conical ground joint neck and comply with ISO 4796-2. The narrow neck is suitable for liquids and the angle of the shoulders ensures that the contents can be smoothly poured. The glass stopper provides an excellent seal that protects the contents from evaporation, oxidation, and contamination. The bottles have a Retrace Code for manufacturing lot traceability and a marking field in permanent white enamel.

  • All borosilicate glass components and therefore suitable for storage of aggressive materials
  • Tight closure due to standard ground joint
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic print

Specifications

Capacity (Metric) 250 mL
Description Neck with standard ground joint and glass flat-headed stopper
For Use With (Application) Storage of air sensitive reagents, pharmaceuticals, and chemicals; Storage of hygroscopic liquids and pharmaceuticals; Transport and shipping of samples; Storage of hygroscopic liquids and pharmaceuticals
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Diameter (Metric) 70 mm
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.