missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Replacement Cap Liner, PTFE-Coated, Peroxide Cured Silicone

Product Code. 11755434 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GL14
GL 18
GL 25
GL 32
GL 45
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) Unit Size
11755434 GL 25 Pack of 10
11381475 GL 45 Pack of 10
11736642 GL14 Pack of 10
11746642 GL 18 Pack of 10
11765434 GL 32 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11755434 Supplier DWK Life Sciences Supplier No. 292481304
  • ONLINE EXCLUSIVE PRICE
    17.35 €
    15.60 € / Pack of 10
    Save 1.75 €
    10% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

DURAN™ Replacement Cap Liner, PTFE-Coated, Peroxide Cured Silicone provides a reliable seal for DURAN™ PBT screw caps. PTFE coating delivers excellent chemical resistance, with heat resistance to 130°C (steam) and 200°C (dry heat). Available in multiple GL thread sizes.

  • Replacement cap liners suitable for DURAN PBT screw caps
  • PTFE coating provides excellent chemical resistance
  • Cap liner forms a tight, secure seal with the bottle
  • Peroxide catalysed silicone construction
  • Heat resistant to 130°C (steam) and 200°C (dry heat)
  • Available in multiple GL thread sizes to suit a range of DURAN bottle formats

Specifications

Color Gray
For Use With (Application) Replacement cap liner for red PBT caps
Material Silicone, PTFE, VMQ
Quantity 10/Pk.
Sterility Non-sterile
Thread Size (Metric) GL 25
Diameter (Metric) 23.5 mm
Autoclavable Autoclavable
Disposable Reusable
Product Type Teflon Liner
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.