Learn More
DWK Life Sciences DURAN™ Screw Thread Coupling, from PBT
Product Code. 10078440 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size:
GL 14
GL 18
GL 25
GL 32
GL 45
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size Unit Size
10078440 GL 32 Each
10383661 GL 14 Each
10768351 GL 18 Each
10120271 GL 25 Each
11629818 GL 45 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10078440 Supplier DWK Life Sciences Supplier No. 292260856
  • ONLINE EXCLUSIVE PRICE
    43.01 €
    38.77 € / Pack of 1
    Save 4.24 €
    9% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The screw thread coupling is used to connect two threaded connections.
The screw thread coupling consists of two threaded caps, which are connected to each other via a flexible hose piece. This component enables a flexible connection of two glass components.
  • For flexible connection of two glass screw-thread connections with integral silicone seal (VQM)
Specifications
Autoclavable Autoclavable
Material PBT
For Use With (Application) Connection of glass threads
Packaging Quantity 1 Ea.
Thread Size GL 32