Learn More
DWK Life Sciences DURAN™ Screw Thread Tube, SVL
Product Code. 15804638 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint No:
SVL 15
SVL 22
SVL 30
SVL 42
Unit Size:
Pack of 10
Pack of 50
Product Code. Standard Taper Joint No unitSize
15804638 SVL 22 Pack of 50
15894628 SVL 15 Pack of 50
15814638 SVL 30 Pack of 50
15844548 SVL 42 Pack of 10
4 options
Product Code. 15804638
Brand: DWK Life Sciences 286266208
Only null left
Add to basket
Add to basket
To attach.
The DURAN™ glass thread tube with SVL thread is a finer thread than the GL thread, which is also used for attaching to glass apparatus. The nominal size refers to the core diameter at the thread.
  • Glass thread tube with SVL thread is a finer thread compared to GL thread
  • The nominal size refers to the core diameter at the thread.
  • Safe and easy sealing and loosening of glass-glass components
  • Grease-free working possible
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50/Pk.
Item Description For attachment to glass apparatuses; Installation in glass apparatus and devices
Joint Size 22 mm
Standard Taper Joint No SVL 22
Thickness (Metric) Wall 1.8 mm