missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Silicone Lid, Stretchable

Product Code. 16688432 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Cyan
Green
Pink
Size:
X-Large
2X-Large
Tamaño de la unidad:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Color Size Tamaño de la unidad
16688432 Green 2X-Large Each
16638432 Pink X-Large Each
16648432 Cyan X-Large Each
16658432 Green X-Large Each
16668432 Pink 2X-Large Each
16678432 Cyan 2X-Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16688432 Proveedor DWK Life Sciences N.º de proveedor 291105137
  • ONLINE EXCLUSIVE PRICE
    7.50 €
    6.74 € / Pack of 1
    Save 0.76 €
    10% Off
In Stock
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

The stretchable silicone lid is used to close round and square vessels.

The closure or covering of vessels is an everyday topic in the laboratory. Often glass plates or aluminum foil are used. From the point of view of environmental friendliness and durability, both variants are rather classified as disadvantageous. The newly developed silicone cover is a good alternative to conventional cover variants and is available individually in three sizes or as a set. Due to its elasticity, the lid can be used for a variety of vessel openings, square or round. The silicone lid has a slightly larger tab that can be used as a writing field. The three colors offered - cyan, pink and green - also allow color identification.

The stretchy DURAN™ silicone lids are resistant to both chemicals and heat. They are recommended for use at temperatures between -40 and +180°C. The DURAN™ lids are manufactured from silicone, which is an autoclavable, and microwave-safe material. However due to the pressure changes that occur during autoclaving the lids may become loose, or tear, as a result they are not recommended for this type of application.

  • For covering and sealing laboratory vessels
  • Stretchable and thus suitable for a large number of vessel openings
  • Available in different colors for clear differentiation and identification
  • Easy to remove with the aid of a flap, both with and without gloves
  • Chemically resistant, heat resistant and reusable

Especificaciones

Color Green
Description Autoclavable (Silicone lid only without glassware)
Diameter (Metric) Outer Aprox. 132 to 147 mm
Material Silicone
For Use With (Application) Covering, Color Coding, Identification
No. per Pack 1
Size 2X-Large
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.