missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Silicone Lid, Stretchable, Set XL-XXL

Product Code. 16628432 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Cyan
Green
Pink
Unit Size:
1 set
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
16628432 Green 1 set
16608432 Pink 1 set
16618432 Cyan 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16628432 Supplier DWK Life Sciences Supplier No. 291100069
  • ONLINE EXCLUSIVE PRICE
    16.36 €
    11.00 € / 1 sets
    Save 5.36 €
    32% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The stretchable silicone lid is used to close round and square vessels.

The closure or covering of vessels is an everyday topic in the laboratory. Often glass plates or aluminum foil are used. From the point of view of environmental friendliness and durability, both variants are rather classified as disadvantageous. The newly developed silicone cover is a good alternative to conventional cover variants and is available individually in three sizes or as a set. Due to its elasticity, the lid can be used for a variety of vessel openings, square or round. The silicone lid has a slightly larger tab that can be used as a writing field. The three colors offered - cyan, pink and green - also allow color identification.

The stretchy DURAN™ silicone lids are resistant to both chemicals and heat. They are recommended for use at temperatures between -40 and +180°C. The DURAN™ lids are manufactured from silicone, which is an autoclavable, and microwave-safe material. However due to the pressure changes that occur during autoclaving the lids may become loose, or tear, as a result they are not recommended for this type of application.

  • For covering and sealing laboratory vessels
  • Stretchable and thus suitable for a large number of vessel openings
  • Available in different colors for clear differentiation and identification
  • Easy to remove with the aid of a flap, both with and without gloves
  • Chemically resistant, heat resistant and reusable

Specifications

Color Green
Description Autoclavable (Silicone lid only without glassware)
Diameter (Metric) Outer Stretch Diameter - XL: 102-120mm. XXL: 132-147mm
Material Silicone
For Use With (Application) Covering, Color Coding, Identification
Includes 1 Piece Each Size (XL and XXL)
No. per Pack 2
Size X-Large and 2X-Large
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.