missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Silicone rubber seals (VMQ), without washer

Product Code. 11860620 Shop All DWK Life Sciences Products
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11860620 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11860620 Supplier DWK Life Sciences Supplier No. 292282501
  • ONLINE EXCLUSIVE PRICE
    89.56 €
    80.96 € / Pack of 10
    Save 8.60 €
    9% Off
View all promotions
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gas washing bottles are used for cleaning gases due to a flow through a suitable liquid.

For gas filtration and cleaning, gas washing bottles are required which consist of a gas distribution tube, a suitable bottle and a gas outlet opening. In this test setup, the gas is forced to flow through a liquid before it is released. The gas distribution pipe is immersed in the liquid for this purpose, the gas interferes through it and impurities of the gas are bound in the liquid. The gas is cleaned or washed so to speak. A filter disk in the gas distribution tube creates air bubbles that are as small as possible and thus form as large a contact area as possible.

  • Drechsel type head
  • With screw-connection system
  • Fired-on, highly durable white print
  • Individual parts can also be ordered separately
  • Easy-to-read scale with graduation marks to estimate the content

Specifications

Material Silicone (VMQ)
Type VMQ Rubber Ring (26 x 42 x 5 mm)
For Use With (Application) Cleaning (washing) of gases with solvents; Purification of gas mixtures; With screw thread for quick exchange of the liquid containing bottles; Filtering
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.