Learn More
DWK Life Sciences DURAN™ Staining Tray
Product Code. 10772742 Shop All DWK Life Sciences Products
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. Quantity Unit Size
10772742 10/Pk. Pack of 10
1 options
Product Code. 10772742 Supplier DWK Life Sciences Supplier No. 213170003
  • ONLINE EXCLUSIVE PRICE
    321.30 €
    279.23 € / Pack of 10
    Save 42.07 €
    13% Off
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The removable staining tray with open bottom allows quick immersion and emptying.
Microscopy is a method of viewing objects that lie outside the resolution of the human eye and can be examined. The objects to be observed are magnified to such an extent that they can be displayed as images. For sample preparation, the objects to be examined are cut into very thin slices. These sections are then applied to an object slide and covered with a cover glass and appropriate solutions. To view the sections under the microscope, the sections on the slide must be filled in a staining trough and stained with an appropriate staining solution.
  • The removable tray has an open bottom to facilitate rapid immersion and drainage, reducing carry-over
  • It is not necessary to take out the slides for drying
  • Allows the slides to be transported from one chamber to another
  • Colouring of up to 10 slides with standard dimensions 76 x 26 mm simultaneously or any width up to 52 mm possible
Specifications
Autoclavable Not Autoclavable
Material Borosilicate Glass (3.3)
For Use With (Application) Staining; Manual staining process; Staining of histological and cytological specimens
No. per Pack 10/Pk.
Quantity 10/Pk.