Learn More
DWK Life Sciences DURAN™ Standard Cone, diamond-ground
Product Code. 15854568 Shop All DWK Life Sciences Products
Click to view available options
Joint Size:
13 mm
17 mm
22 mm
26 mm
Unit Size:
Pack of 50
Product Code. 15854568
Brand: DWK Life Sciences 286156303
Only null left
Add to basket
Add to basket
Designed for a rigid, self-supporting connection with lower requirements.
The conical ground joint of sleeve and core is a rigid, self-supporting joint. DURAN™ DG cores are not ground according to DIN 12249 and represent the cost-effective alternative to DURAN™ cores. Ground joints with ground grease or PTFE sleeves should always be used. KECK™ Conical ground joint clamps are the perfect supplement for securing the conical ground joint.
  • Diamond cut (coarser cut quality, more roughness depth)
  • Cost-effective alternative to DURAN™ cores (not according to DIN)
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50/Pk.
Item Description Installation in glass apparatus and devices
Joint Size 22 mm
Thickness (Metric) Wall 1.8 mm