Learn More
DWK Life Sciences DURAN™ Standard Cone, finished
Product Code. 15951531 Shop All DWK Life Sciences Products
Click to view available options
Joint Size:
4 mm
6 mm
8 mm
11 mm
13 mm
17 mm
22 mm
26 mm
30 mm
36 mm
40 mm
46 mm
50 mm
56 mm
65 mm
80 mm
90 mm
Unit Size:
Each
Pack of 10
Pack of 50
Product Code. 15951531
Brand: DWK Life Sciences 286153203
Only null left
Add to basket
Add to basket
For a rigid, self-supporting connection.
The conical ground joint is the most common joint in glass apparatus engineering. Ground DURAN™ cores comply with DIN 12249 Form B in their design and meet higher requirements for surface roughness. This reduces the need for grinding grease and the risk of jamming. Ground joints with ground grease or PTFE sleeves should always be used. KECK™ Conical ground joint clamps are the perfect supplement for securing the conical ground joint.
  • According to DIN 12249 Form B
  • Ground (finer grinding quality, less roughness depth)
  • The finer surface reduces the risk of jamming the cuts.
  • Minimized grease requirement
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50/Pk.
Item Description Installation in glass apparatus and devices
Joint Size 13 mm
Thickness (Metric) Wall 1.5 mm