missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Duran™ 'T' Shaped Tubing Connectors

Brand:  Lenz Laborglasinstrumente™ 01600508

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10048470

  • 4.45 € / Each
Estimated Shipment: 22-01-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Tubing Connector
Autoclavable
115 mm
T
8 mm
Duran Glass
1 Pack
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers