Learn More
DWK Life Sciences DURAN™ TILT Bottle Light Shield, White, Silicone Rubber
Product Code. 15593537 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 4
Product Code. Color Unit Size
15593537 White Pack of 4
1 options
Product Code. 15593537 Supplier DWK Life Sciences Supplier No. 292435601
  • ONLINE EXCLUSIVE PRICE
    37.90 €
    37.27 € / Pack of 4
    Save 0.63 €
    1% Off
Estimated Shipment: 29-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The DURAN™ TILT silicone sleeve blocks damaging UV light, and facilitates safer handling.
The DURAN™ TILT Light Shield is a white silicone sleeve that covers 94% of the bottle surface. The sleeve has a number of protective functions: it blocks damaging ultraviolet light (UV), protects the glass surface from damage, and facilitates safer handling. DURAN™ TILT Light Shield includes four GL 56 Bottle Tags (Orange, Yellow, Blue, Purple) made from silicone.
  • The white DURAN™ TILT Light Shield is a silicone sleeve that covers 94 % of the bottle surface and blocks damaging ultraviolet light
  • Protects the glass surface from damage
  • Facilitates safer handling
  • Silicone provides a much better grip than glass especially when the bottle has been sprayed with 70 % alcohol for surface disinfection
Specifications
Color White
Material Silicone
For Use With (Application) Working with cell culture media under sterile conditions in biosafety cabinets and clean hoods; Storage of light-sensitive cell culture media
No. per Pack 4/Pk.
Autoclavable Autoclavable