Learn More
DWK Life Sciences DURAN™ Tube with Screw Thread, with DIN thread
Product Code. 13167613 Shop All DWK Life Sciences Products
Click to view available options
Thread Size:
GL 14
GL 18
GL 25
GL 32
GL 45
Unit Size:
Each
Pack of 10
Product Code. 13167613
Brand: DWK Life Sciences 248350103
Only null left
Add to basket
Add to basket
The threaded tube is used for further processing in glass components.
This threaded tube has a screw thread and can be incorporated into glass components by a further processing step.
  • With DIN thread
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (3.3)
For Use With Shoring in glass apparatuses
Quantity 1 Ea.
Thread Size GL 45
Autoclavable Autoclavable
Diameter (Metric) 40 mm
Wall Thickness 2.3 mm