missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Weighing Bottles

Product Code. 10748841
Change view
Click to view available options
Quantity:
6Pack
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10748841 6Pack Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10748841 Supplier Lenz Laborglasinstrumente™ Supplier No. 05560019
  • ONLINE EXCLUSIVE PRICE
    152.00 €
    100.23 € / Pack of 6
    Save 51.77 €
    34% Off
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the precise and secure weighing of samples in the laboratory. Lenz Laborglasinstrumente™ Weighing Bottles are manufactured from durable DURAN™ borosilicate glass.

  • With interchangeable NS stopper or cap
  • With inscription label
  • With fused-in size (mm) and capacity (mL)
  • Available in high- or low-forms
  • Corrosion-free and resistant to thermal shock

Specifications

Form Tall
Includes Interchangeable NS stopper, inscription label
Quantity 6Pack
Stopper Number NS 40/12
Product Line Bottle
Height (Metric) 65 mm
Product Type Weighing Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.