missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ Durete Reagent Refills

Product Code. 11938067
Change view
Click to view available options
Quantity:
250/Pk.
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11938067 250/Pk. Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11938067 Supplier Palintest™ Supplier No. AP254
  • ONLINE EXCLUSIVE PRICE
    177.80 €
    126.04 € / Pack of 250
    Save 51.76 €
    29% Off
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Robust and reliable durete reagent refills for photometers.

  • X250 refill photom durete 0-500PPM
  • Durete Reagent Refill
  • Hardness, Photometer Reagents, 0 - 500 mg/L CaCO3, (50/pk)

Specifications

Concentration 0 to 500ppm
Packaging 250/Pk.
Quantity 250/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.