missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dutasteride, 99%

Product Code. 15618745 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15618745 250 mg 250mg
15608745 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15618745 Supplier Thermo Scientific Chemicals Supplier No. 461512500
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 164656-23-9
Molecular Formula C27H30F6N2O2
Molecular Weight (g/mol) 528.53
InChI Key JWJOTENAMICLJG-QWBYCMEYSA-N
Synonym dutasteride, avodart, avolve, duagen, unii-o0j6xjn02i, avodart tn, o0j6xjn02i, dutasteride jan/usan/inn, 5alpha,17beta-n-2,5-bis trifluoromethyl phenyl-3-oxo-4-azaandrost-1-ene-17-carboxamide, dutasteride usan
PubChem CID 6918296
ChEBI CHEBI:521033
IUPAC Name (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-[2,5-bis(trifluoromethyl)phenyl]-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide
SMILES CC12CCC3C(C1CCC2C(=O)NC4=C(C=CC(=C4)C(F)(F)F)C(F)(F)F)CCC5C3(C=CC(=O)N5)C

Specifications

Melting Point 245.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: insoluble.
Formula Weight 528.53
Percent Purity 99%
Chemical Name or Material Dutasteride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.