missing translation for 'onlineSavingsMsg'
Learn More

Dutasteride, TRC

Product Code. 18001040
Change view
Click to view available options
Quantity:
5 mg
100 mg
1 g
Unit Size:
100mg
1g
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18001040 1 g 1g
18032830 5 mg 5mg
18050740 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18001040 Supplier Toronto Research Chemicals Supplier No. TRCD7350001G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 164656-23-9
Molecular Formula C27 H30 F6 N2 O2
Molecular Weight (g/mol) 528.53
Synonym 1H-Indeno[5,4-f]quinoline-7-carboxamide, N-[2,5-bis(trifluoromethyl)phenyl]-2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-, 4-Azaandrost-1-ene-17-carboxamide, N-[2,5-bis(trifluoromethyl)phenyl]-3-oxo-, (5α,17β)-, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-N-[2,5-Bis(trifluoromethyl)phenyl]-2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-1H-indeno[5,4-f]quinoline-7-carboxamide, (5α,17β)-N-[(2,5-Bis(trifluoromethyl)phenyl]-3-oxo-4-aza-5-androst-1-ene-17-carboxamide, Avodart, Avolve, Dutasteride, GG 745, GI 198745, Veltride
IUPAC Name (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-[2,5-bis(trifluoromethyl)phenyl]-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide
SMILES C[C@]12CC[C@H]3[C@@H](CC[C@H]4NC(=O)C=C[C@]34C)[C@@H]1CC[C@@H]2C(=O)Nc5cc(ccc5C(F)(F)F)C(F)(F)F

Specifications

Quantity 1 g
InChI Formula InChI=1S/C27H30F6N2O2/c1-24-11-9-17-15(4-8-21-25(17,2)12-10-22(36)35-21)16(24)6-7-19(24)23(37)34-20-13-14(26(28,29)30)3-5-18(20)27(31,32)33/h3,5,10,12-13,15-17,19,21H,4,6-9,11H2,1-2H3,(H,34,37)(H,35,36)/t15-,16-,17-,19+,21+,24-,25+/m0/s1
Formula Weight 528.2211
Percent Purity >95%
Chemical Name or Material Dutasteride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.