missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Dye Tablets
Shop All Cole-Parmer Products
Click to view available options
Color:
Dark Red
Yellow/Green
Unit Size:
Pack of 200
Description
Cole-Parmer Dye Tablets, Yellow/Green, Dye Tracer, For Use With: Plumbing, storm and sanitary drain analysis, Quantity: 200/Pk.
Specifications
Specifications
| Color | Dark Red |
| For Use With (Application) | Plumbing, storm and sanitary drain analysis |
| Quantity | 200/Pk. |
| Type | Dye Tracer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction