missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyLight™ 680 Microscale Antibody Labeling Kit

Product Code. 10016763 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 Reactions
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10016763 5 Reactions 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10016763 Supplier Thermo Scientific™ Supplier No. 53057

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Label and purify antibodies for fluorescence imaging assays in one hour with the full spectral range of activated DyLight Fluorescent Dye kits.

Thermo Scientific™ DyLight Fluor Antibody Labeling Kits provide the essential reagents and accessories to easily label and purify antibodies and include regular-scale and microscale kits for each specific DyLight Fluorescent Dye.

Two convenient kit formats are available to accommodate varied labeling requirements. The DyLight Antibody Labeling Kits contain all necessary components to perform three separate labeling reactions using 1mg of IgG or similar quantities of other proteins. The DyLight Microscale Antibody Labeling Kits contain all the necessary components to perform five separate labeling reactions using 100μg of IgG. The labeling kits use high-performance spin desalting columns to provide exceptional dye removal and antibody recovery.

Highlights:

  • Fast - label and purify protein in approximately one hour
  • Amine-reactive dyes - label virtually any protein
  • Efficient non-reacted dye removal
  • Minimal sample dilution
  • Spin column format eliminates the need for column preparation, fraction screening and waiting for protein to emerge from column

Item-Specific Features:

  • Standard kits contain sufficient reagents to label and purify 3 × 1mg of IgG or similar quantities of other proteins
  • Microscale kits contain sufficient reagents to label and purify 5 × 0.1mg of IgG or similar quantities of other proteins

Includes:

Antibody Labeling Kits

DyLight NHS Ester, 3 vials; Reaction buffer, 1mL; Spin columns, 6 each; Microcentrifuge collection tubes, 12 each; Purification resin, 5mL

Microscale Kits

DyLight NHS Ester, 5 vials; Reaction buffer, 1mL; Spin columns, 5 each; Microcentrifuge collection tubes, 10 each; Purification resin, 5mL

TRUSTED_SUSTAINABILITY

Specifications

Description DyLight 680 Antibody Labeling Microscale Kit
Product Type Antibody Labeling Kit
Quantity 5 Reactions
Sufficient For Labeling and purifying 5 x 100 μg (1 mg/mL) of IgG or other proteins
Content And Storage Sufficient For: Labeling and purifying 5 x 100 μg (1 mg/mL) of IgG or other proteins
  • DyLight Fluor NHS Ester, 5 x 15 μg vials
  • Borate Buffer (0.67M), 1 mL
  • Purification Resin, 5 mL
  • Microcentrifuge Spin Columns, 5 columns
  • Microcentrifuge Collection Tubes, 10 tubes

    Store the DyLight NHS Ester at -20°C. Store all other kit components at 4°C.
  • Excitation/Emission 692/712
    Labeling Scale 100 μg
    Product Line DyLight
    Label or Dye DyLight 680
    Labeling Target Antibodies, Proteins
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.