missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyLight™ Antibody Labeling Kits

Product Code. 10381064 Shop All Thermo Scientific Products
Change view
Click to view available options
Excitation/Emission:
346/442
400/420
490/525
562/576
590/617
638/658
652/672
679/702
749/775
784/814
Label or Dye:
DyLight 350
DyLight 405
DyLight 488
DyLight 550
DyLight 594
DyLight 633
DyLight 650
DyLight 680
DyLight 755
DyLight 800
Unit Size:
1 set
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Excitation/Emission Label or Dye Unit Size
10381064 490/525 DyLight 488 1 set
10741955 400/420 DyLight 405 1 set
10197873 590/617 DyLight 594 1 set
10391404 638/658 DyLight 633 1 set
10270154 679/702 DyLight 680 1 set
10026763 784/814 DyLight 800 1 set
11891184 346/442 DyLight 350 1 set
11814574 562/576 DyLight 550 1 set
11834574 652/672 DyLight 650 1 set
11572501 749/775 DyLight 755 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 10381064 Supplier Thermo Scientific™ Supplier No. 53024
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Covalently label antibodies or proteins with DyLight dyes for use in immunofluorescence, flow cytometry, western blot analysis, and high content analysis.

Invitrogen Dylight Antibody Labeling kits provide everything needed to label 1 mg of antibody or protein with amine-reactive Dylight dye and subsequent free dye removal. Choose from a variety of Dylight fluorophores across the spectrum to covalently label your antibody or protein. Labeled antibodies are typically ready to use in 90 minutes with minimal hands-on time. The kit contains all necessary components to perform three separate labeling reactions using 1 mg of IgG or similar quantities of other proteins.

Invitrogen DyLight Antibody Labeling kits enable the creation of fluorescent antibodies that are covalently linked to different DyLight fluorophores. The fluorescently labeled antibodies can be used in a wide variety of laboratory applications, including immunofluorescence microscopy, IHC, western blotting, ELISA, flow cytometry, or in vitro and in vivo fluorescent detection strategies.

DyLight Antibody Labeling kits contain an NHS ester-activated derivative of the high-performance DyLight dye that can be used to fluorescently label antibodies and other proteins for cellular imaging and other fluorescence detection methods. The DyLight dye is an amine-reactive dye and is activated with an N-hydroxysuccinimide (NHS) ester moiety to react with exposed N-terminal α-amino groups or the ε-amino groups of lysine residues to form stable amide bonds.

DyLight Antibody Labeling kits contain all necessary components to perform three separate labeling reactions and free dye removal using 1 mg of IgG or similar quantities of other proteins.

DyLight dyes exhibit higher fluorescence intensity and photostability than CyDye and LI-COR™ dyes in many applications and remain highly fluorescent over a broad pH range (pH 4–9). Additionally, the water solubility of the DyLight fluorescent dyes allows a high dye-to-protein ratio to be achieved without causing precipitation of conjugates. Finally, the high solubility of these fluorophores permits protein solutions to be added directly to specific amounts of the labeling reagent.

Other advantages of the DyLight Antibody Labeling kits:
· High performance—high fluorescence intensity across a broad pH range
· Multiple colors—10 different dyes across the visible spectrum and extending into the infrared
· Specificity—NHS ester-activated dye labels proteins and other molecules at primary amines (-NH2)
· Optimized procedure—following the standard protocol results in antibodies with excellent dye:protein ratios and recovery rates for optimum activity and fluorescence labeling

We also offer DyLight Microscale Antibody Labeling kits for fast and efficient fluorescent labeling of antibodies for use in fluorescence methods. The DyLight Microscale Antibody Labeling kits contain all necessary components to perform five separate labeling reactions using 100 μg of IgG or similar quantities of other proteins. Both the standard and microscale kit sizes include the amine-reactive DyLight fluorophore in convenient single-use vials as well as purification resin and spin columns for the preparation of ready-to-use conjugate.
TRUSTED_SUSTAINABILITY

Specifications

Kit Contents Contains DyLight Fluor NHS Ester, 5 x 15 μg vials; Borate Buffer (0.67 M), 1 mL; Purification Resin, 5 mL; Microcentrifuge Spin Columns, 5 columns; Microcentrifuge Collection Tubes, 10 Tubes
Product Type Labeling Kit
Quantity 3 reactions
Content And Storage Store the DyLight NHS Ester at -20°C. Store all other kit components at 4°C.
Excitation/Emission 490/525
Labeling Scale 0.5 to 1 mg
Product Line DyLight
Chemical Reactivity Amine
Label or Dye DyLight 488
Color Green
Labeling Target Antibodies, Proteins
Label Type DyLight™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.