missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Dynabeads™ CaptureSelect™ AAV9 Magnetic Beads

Product Code. 30276152 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
50 mL
Unit Size:
1mL
50mL
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30276152 50 mL 50mL
30275957 1 mL 1mL
30276138 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30276152 Supplier Thermo Scientific™ Supplier No. 2853332050
  • ONLINE EXCLUSIVE PRICE
    8405.00 €
    8290.00 € / 50mL
    Save 115.00 €
    1% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Dynabeads™ CaptureSelect™ AAV9 Magnetic Beads are high-capacity, high-throughput magnetic affinity particles for the purification of naturally occurring and synthetic adeno-associated virus serotype 9 (AAV9) used for gene therapy applications.

Dynabeads CaptureSelect AAV9 Magnetic Beads are high-capacity, high-throughput magnetic affinity particles for the purification of naturally occurring and synthetic adeno-associated virus serotype 9 (AAV9) used for gene therapy applications. The anti-AAV9 affinity ligand demonstrates good binding reactivity towards the AAV9 serotype. The magnetic beads enable purification using manual and robotic magnetic separators of the AAV9 serotype directly from crude feedstock in a single step with high purity and yield.

Features of DynaBeads CaptureSelect AAV9 Magnetic Beads include:

  • One-magnetic-step AAV purification from crude material with high purity and yield
  • Compatible with manual and automated workflows
  • High specificity and capacity
  • Easy upscaling to higher volumes
  • Non-animal derived

The magnetic beads bind with high affinity specifically to the AAV9 serotype. For the isolation of other AAV serotype capsids we recommend DynaBeads CaptureSelect AAVX magnetic beads (Cat. No. 2853522001, 2853522005, 2853522050).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a ∼12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain. The ligand is efficiently produced in yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Bead matrix: non-porous monodisperse superparamagnetic beads
  • Bead size: 4.5 μm
  • Ligand: CaptureSelect AAV9 affinity ligand
  • Binding capacity: >5 x 1011 viral particles/mg beads
  • Elution conditions: 50 mM citric acid, pH 2.5–3.0
  • Formulation buffer: tris-buffered saline, 0.05% Tween 20, 0.05 % sodium azide

Specifications

Concentration 30 mg beads/mL slurry, 1 mg contains approximately 1.3 x 107 beads
Formulation 4 to 6% (v/v) slurry in Tris buffered saline with 0.05% Tween 20 and 0.05% sodium azide
Matrix Non-porous monodisperse superparamagnetic beads
Content And Storage Store at 2°C to 8°C. Do not freeze.
Shipping Condition Wet Ice
Particle Size 4.5 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 50 mL
For Use With (Application) Magnetic isolation of AAV9 capsids
Target AAV9 (Adeno-associated serotype 9)
Type Magnetic Affinity Bead
Ligand Type CaptureSelect AAV9 Affinity Ligand
Sufficient For Sample Preparation
Binding Capacity >0.5 x 1011 vp/mg Beads
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.