missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ In Vitro Transcription and RNA Purification Kit

Product Code. 30055979
Change view
Click to view available options
Quantity:
2 mL
10 mL
Size:
Large Combo Kit
Small Combo Kit
Unit Size:
10mL
2mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Size unitSize
30055979 2 mL Small Combo Kit 2mL
30056113 10 mL Large Combo Kit 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30055979 Supplier Invitrogen™ Supplier No. 65034D
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The Dynabeads In Vitro Transcription and RNA Purification Kit offers an innovative technology for simple, flexible, and automation-ready mRNA production and is particularly suitable where high quality and scalability are required.

The Dynabeads In Vitro Transcription and RNA Purification Kit offers an innovative technology for simple, flexible, and automatable mRNA production and is particularly suitable where high quality and scalability are required, such as in the discovery phase of RNA-based vaccines and therapeutics development. This automation-ready solution can help speed up the screening of hundreds of constructs that may be required to identify a final drug candidate, in scalable quantities to match your requirements.

Features of the Dynabeads In Vitro Transcription and RNA Purification Kit include:

  • Flexibility – choose DNA template for IVT with flexibility of design and amplification
  • Efficiency – reuse DNA template and beads (at least six times) to help reduce scost and eliminate/minimize plasmid prep hands-on time and risk of bacterial host DNA transfer
  • Scalability – swap between manual and automated workflows to match required scale (micrograms to grams of mRNA) and throughput
  • Quality – help ensure a safe and flexible supply of magnetic beads to navigate from RNA-based drug discovery research to commercial manufacturing with counterpart solutions (available via the OEM commercial supply), without incurring extra time or costs from switching technology or raw material

Overview of the workflow
A biotinylated template is made by PCR amplification of the target sequence in a plasmid construct or in a synthetic DNA construct using a biotinylated forward primer and a non-biotinylated reverse primer (figure below). As an alternative to PCR amplification, a linearized plasmid can be biotinylated by a fill-in reaction of biotin-dUTP in the 5' overhang sequence, provided the correct plasmid design. The biotinylated template is then immobilized directly to the included Dynabeads Streptavidin for In Vitro Transcription beads without the need for prior purification. The bead-bound template is used directly in the IVT, then removed from the synthesized mRNA by magnetic separation. The template can be reused (at least six times) in consecutive IVT reactions (figure below).

Solid-phase IVT is compatible with commonly used reagents such as the MEGAscript T7 Transcription Kit, mMESSAGE mMACHINE T7 Transcription Kit, and mMESSAGE mMACHINE T7 ULTRA Transcription Kit, based on your needs and strategies for capping and polyadenylation.

Crude RNA from the IVT step is mixed with the included Dynabeads Carboxylic Acid for RNA Purification and the included Dynabeads RNA Binding Buffer is added. The RNA binds to the bead surface and the remaining components of the reaction mixture are removed by applying a magnet (see thermofisher.com/magnets) and discarding the supernatant (figure below).

Automation-ready for use with KingFisher purification systems
The 1-μm magnetic beads are ideal for high-throughput screening. The process can be automated using liquid handlers or any of our KingFisher sample purification systems. Automation with a KingFisher instrument enables IVT template immobilization, IVT reaction, and purification of 96 samples in less than four hours, and up to 5 mg mRNA yield per well. The scripts for KingFisher Flex and KingFisher Apex automated purification workflows are available from our website.

Commercial supply
To learn more about counterpart products and support documentation available for GMP-compliant manufacturing of mRNA vaccines and therapeutics, visit: thermofisher.com/dynabeadsmrna.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 unit of Dynabeads Streptavidin for In Vitro Transcription (Cat. No. 65005D). Store at 2–8°C.
• 1 unit of Dynabeads Carboxylic Acid for RNA Purification (Cat. No. 65020D). Store at 2–8°C.
• 1 unit of Dynabeads RNA Binding Buffer (Cat. No. 65040D). Store at 2–8°C. Protect from light exposure during storage.
Description Small Combo Kit
Isolation Technology Magnetic Bead
Purification Time Variable
For Use With (Equipment) KingFisher™ Sample Purification System, DynaMag™ magnets
For Use With (Application) In Vitro Transcription, Synthetic RNA, Crude RNA Purification (from IVT Reaction Mix)
Product Line Dynabeads
Quantity 2 mL
RNase RNase-free (No detectable activity level)
Shipping Condition Ambient Temperature
Size Small Combo Kit
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.