missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ RNA Binding Buffer

Product Code. 17847378
Change view
Click to view available options
Quantity:
20 mL
50 mL
Unit Size:
20mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17847378 20 mL 20mL
17804058 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 17847378 Supplier Invitrogen™ Supplier No. 65040D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Dynabeads RNA Purification Buffer is intended for use with Dynabeads Carboxylic Acid for RNA Purification, providing a simple, scalable, and automation-ready solution for purification of crude RNA produced by in vitro transcription (IVT).

Dynabeads RNA Binding Buffer is intended for use with Dynabeads Carboxylic Acid for RNA Purification beads, together providing a simple, scalable, and automatable solution for purification of crude RNA produced by in vitro transcription (IVT). The products may be used for purification following solid-phase IVT with Dynabeads Streptavidin for In Vitro Transcription (figure below). The combination is particularly suitable where flexibility and high purity are required, such as in the discovery phase of RNA-based vaccines and therapeutics development.

Features of Dynabeads RNA Binding Buffer
This buffer, combined with Dynabeads Carboxylic Acid for RNA Purification, offers efficiency, scalability (micrograms to grams) in manual and automated workflows, and quality to navigate from RNA-based drug discovery to commercial manufacturing without incurring extra time or costs from switching technology or raw material.

The buffer is offered as a stand-alone product. It is also included in the Dynabeads RNA Purification and Dynabeads In Vitro Transcription and RNA Purification kits.

Overview of the workflow
Crude RNA from an IVT reaction is mixed with the beads (not included) and the buffer is added. The RNA binds to the bead surface and the remaining components of the reaction mixture are removed by applying a magnet (see thermofisher.com/magnets) and discarding the supernatant (figure below).

Automation-ready for use with KingFisher purification systems
The 1-μm magnetic beads are ideal for high-throughput screening. The process can be automated using liquid handlers or any of our KingFisher sample purification systems. The scripts for KingFisher Flex and KingFisher Apex automated purification workflows are available from our website.

Commercial supply
To learn more about counterpart products and support documentation available for GMP-compliant manufacturing of mRNA vaccines and therapeutics, visit: thermofisher.com/dynabeadsmrna.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2°C to 8°C and protected from light.
Isolation Technology Magnetic Bead
Purification Time Variable
Sample Type Crude RNA
For Use With (Equipment) KingFisher™ Sample Purification System, DynaMag™ magnets
For Use With (Application) In Vitro Transcription, Synthetic RNA, Crude RNA Purification (from IVT Reaction Mix)
Product Line Dynabeads
Quantity 20 mL
RNase No detectable activity level
Shipping Condition Ambient Temperature
Target RNA
Volume (Metric) 20 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.