missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ RNA Purification Kit

Product Code. 30055080
Change view
Click to view available options
Quantity:
2 mL
10 mL
Unit Size:
10mL
2mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30055080 10 mL 10mL
30056333 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30055080 Supplier Invitrogen™ Supplier No. 65032D
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Dynabeads RNA Purification Kit contains Dynabeads RNA Purification Buffer and Dynabeads Carboxylic Acid for RNA Purification, providing a simple, flexible, and automation-ready solution for purification of crude RNA produced by in vitro transcription (IVT).

The Dynabeads RNA Purification Kit contains Dynabeads RNA Binding Buffer and Dynabeads Carboxylic Acid for RNA Purification beads, providing a simple, flexible, and automatable solution for purification of crude RNA produced by in vitro transcription (IVT). It is particularly suitable where high purity and scalability are required, such as in the discovery phase of RNA-based vaccines and therapeutics development.

Features of the Dynabeads RNA Purification Kit include:

  • Efficiency – recycle (at least six times) to help reduce the cost and time in simple workflow
  • Scalability – swap between manual and automated workflows to match required scale (micrograms to grams of mRNA) and throughput
  • Quality – help ensure a safe and flexible supply of products to navigate from RNA-based drug discovery research to commercial manufacturing with counterpart solutions (available via the OEM commercial supply) without incurring extra time or costs from switching technology or raw material

Overview of the workflow
Crude RNA from the IVT step is mixed with the Dynabeads Carboxylic Acid for RNA Purification and Dynabeads RNA Binding Buffer is added. The RNA binds to the bead surface and the remaining components of the reaction mixture are removed by applying a magnet (see thermofisher.com/magnets) and discarding the supernatant (figure below).

The products offered in this combo kit can also be ordered separately. In addition, they are included in the Dynabeads In Vitro Transcription and RNA Purification Kit along with Dynabeads Streptavidin for In Vitro Transcription.

Automation-ready for use with KingFisher purification systems
The 1-μm magnetic beads are ideal for high-throughput screening. The process can be automated using liquid handlers or any of our KingFisher sample purification systems. The scripts for KingFisher Flex and KingFisher Apex automated purification workflows are available from our website.

Commercial supply
To learn more about counterpart products and support documentation available for GMP-compliant manufacturing of mRNA vaccines and therapeutics, visit: thermofisher.com/dynabeadsmrna.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 unit of Dynabeads Carboxylic Acid for RNA Purification (Cat. No. 65021D). Store at 2–8°C.
• 1 unit of Dynabeads RNA Binding Buffer (Cat. No. 65042D). Store at 2–8°C. Protect from light exposure during storage.
Description Large Combo Kit
Isolation Technology Magnetic Bead
Purification Time Variable
Sample Type Crude RNA
High-throughput Compatibility Yes
For Use With (Equipment) KingFisher™ Sample Purification System, DynaMag™ magnets
For Use With (Application) In Vitro Transcription, Synthetic RNA, Crude RNA Purification (from IVT Reaction Mix)
Product Line Dynabeads
Quantity 10 mL
RNase RNase-free (No detectable activity level)
Shipping Condition Ambient Temperature
Size Large Combo Kit
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.