missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DynaMag™-50 Magnet

Product Code. 10469193
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10469193 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10469193 Supplier Invitrogen™ Supplier No. 12302D
Curiosity Goes Beyond
Use promo code 27966
View all promotions
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

The DynaMag-50 magnet combines a strong magnetic attraction for efficient separations with a flexible and ergonomic design that ensures comfortable working position.

The DynaMag-50 magnet combines a strong magnetic attraction for efficient separations with a flexible and ergonomic design that ensures comfortable working position. A tilting mechanism on magnet enables you to rest your elbows while pipetting. The magnet can be used for any starting sample in combination with all types of cell-isolation Dynabeads™.

  • Optimal working volume: 5—50mL
  • Magnet holds 2 x 50 mL tubes
  • Excellent control and visibility of your isolations

This magnet replaces the Dynal MPC™-50 magnet

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Equipment) DynaMag™
Shipping Condition Room Temperature
Product Line DynaMag
Quantity 1 Each
Type Cell Separation Magnet
Sample Volume 10-50 mL
What are the Gauss levels for the DynaMag-2 and DynaMag-Spin Magnets?

The DynaMag-2 Magnet is 3,500-3,700 minimum Gauss level and the DynaMag-Spin is 3,000-3,500 minimum Gauss level.

What is the standard conversion of Gauss to Tesla units?

1 Tesla is equivalent to 10,000 Gauss. .

How can I clean my DynaMag magnet?

Spray and/or wipe the DynaMag magnet unit with one of the following cleaning agents:

- 70% (v/v) isopropyl alcohol
- 1% (v/v) sodium hypochlorite solution (bleach)
- 0.1 N HCl solution

Do not submerge the magnet unit in aqueous solutions and avoid prolonged exposure to water or aqueous solutions. Clean with a damp cloth and mild detergent when exposed to harsh solvents. Do not autoclave the magnet unit as heating the magnet to above 70 degrees C can destabilize the magnetic material, resulting in loss of magnetic strength.

What is the difference between the DynaMag-2 Magnet (Cat. No. 12321D) and DynaMag-Spin Magnet (Cat. No. 12320D)?

The DynaMag-2 Magnet can hold 16 standard 1.5 mL or 2 mL microcentrifuge tubes, while the DynaMag-Spin Magnet holds up to 6 standard 1.5 mL microcentrifuge tubes.

What is the definition of superparamagnetic, and what does this mean for my cell isolation application with Dynabeads magnetic beads?

Superparamagnetic means that the Dynabeads magnetic beads exhibit magnetic properties when placed within a magnetic field, but have no residual magnetism when removed from the magnetic field.

This means that your targeted cells, proteins, or nucleic acids are only subjected to magnetic forces during the time the beads are on the magnet. The beads do not aggregate, but remain evenly dispersed in suspension.

Are the antibodies on your Dynabeads magnetic beads for cell isolation/activation/expansion covalently bound to the beads?

Yes. The antibodies are covalently bound and should be very stable in your applications.

What is the average shelf life of Dynabeads magnetic beads?

Depending on the antibody coated on the Dynabeads magnetic beads, the shelf life can vary from 24-36 months.
Some kits may have 18 months shelf life depending on other components supplied in the kit. The kits are guaranteed for 6 months from when you receive them. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

What is the strength of the DynaMag-50 Magnet (Cat. No. 12302D)?

The strength of the DynaMag-50 Magnet is a minimum of 4,250 Gauss.

How can I clean my DynaMag magnet?

Spray and/or wipe the DynaMag magnet unit with one of the following cleaning agents:

- 70% (v/v) isopropyl alcohol
- 1% (v/v) sodium hypochlorite solution (bleach)
- 0.1 N HCl solution

Do not submerge the magnet unit in aqueous solutions and avoid prolonged exposure to water or aqueous solutions. Clean with a damp cloth and mild detergent when exposed to harsh solvents. Do not autoclave the magnet unit as heating the magnet to above 70 degrees C can destabilize the magnetic material, resulting in loss of magnetic strength.

How can I clean my DynaMag magnet?

Spray and/or wipe the DynaMag magnet unit with one of the following cleaning agents:

- 70% (v/v) isopropyl alcohol
- 1% (v/v) sodium hypochlorite solution (bleach)
- 0.1 N HCl solution

Do not submerge the magnet unit in aqueous solutions and avoid prolonged exposure to water or aqueous solutions. Clean with a damp cloth and mild detergent when exposed to harsh solvents. Do not autoclave the magnet unit as heating the magnet to above 70 degrees C can destabilize the magnetic material, resulting in loss of magnetic strength.

Which magnet should I use with my Dynabeads magnetic beads?

We recommend that you select a magnet based on your sample volume:

- Invitrogen DynaMag-2 Magnet (Cat. No. 12321D) magnet working volume: 1 µl-1.5 mL sample volume
- Invitrogen DynaMag-5 Magnet (Cat. No. 12303D) magnet working volume: 1-5 mL sample volume
- Invitrogen DynaMag-15 Magnet (Cat. No. 12301D) magnet working volume: 4-15 mL sample volume
- Invitrogen DynaMag-50 Magnet (Cat. No. 12302D) magnet working volume: 10-50 mL sample volume
- Invitrogen DynaMag-96 Side Magnet (Cat. No. 123-31D): pulls beads to the side of each well. Compatible with PCR-strips and 96-well plates (200µL), non-skirted and half-skirted
- Invitrogen DynaMag-96 Bottom Magnet (Cat. No. 123-32D): pulls beads to the bottom of each well. Compatible with PCR-strips and 96-well plates (200µL) non-skirted and half-skirted


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.