missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ DynaLight™ Trigger Solution

Product Code. 12807452
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12807452 1 L Each
12887442 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12807452 Supplier Molecular Probes™ Supplier No. 4475409
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Achieve faster ELISA results with DynaLight™ Trigger Solution when used in combination with DynaLight™ Substrate with RapidGlow™ Enhancer, a ready-to-use chemiluminescent substrate formulation that has been optimized to achieve faster results in solution-based assays.

Features of DynaLight™ Substrate with RapidGlow™ Enhancer:

• Faster kinetics and higher signal levels than colorimetric, fluorescent, or other chemiluminescent chemistries
• Sensitive attomole detection of purified alkaline phosphatase enzyme
• Optimized formulation for both microplate and bead-based assays
• Substrate stability of over 1 year at 4°C
• Compatible with automated platforms with trigger solution/positions already in place

The DynaLight™ Substrate with RapidGlow™ Enhancer provides a robust chemiluminescent reaction when catalyzed by alkaline phosphatase. This reaction achieves maximum light emission within 2 minutes and maintains a sustained signal for hours. When combined with DynaLight™ Trigger Solution, DynaLight™ Substrate with RapidGlow™ Enhancer delivers maximum signal within seconds. The environmentally-friendly DynaLight™ Trigger Solution provides maximum signal within 1 second of addition, with a longer half-life to its ground state than other chemiluminescent Flash technologies, allowing ample time to collect data reliably.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Trigger Solution
Quantity 1 L
Detection Method Chemiluminescence
Assay Sensitivity 1 pg/mL
Form Liquid
For Use With (Equipment) Luminometer (Tube-Based)
High-throughput Compatibility High-throughput Compatible
Product Line DynaLight
Sample Type Cell Cultures, Whole Blood, Serum
Substrate Properties Chemical Substrate
Substrate Type Alkaline Phosphatase
Test Time 31 sec.
Content And Storage 2 clear bottle. Store (at room temperature.)
Shipping Condition Room Temperature
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.