Learn More
ABEBA™ Dynamic Anthracite Occupational Clogs Ob ESD
Product Code. 17191355
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17191355 47 1 pair
17171345 35 1 pair
17181345 36 1 pair
17171355 45 1 pair
17181355 46 1 pair
17101365 48 1 pair
17191345 37 1 pair
17101355 38 1 pair
17111355 39 1 pair
17121355 40 1 pair
17131355 41 1 pair
17141355 42 1 pair
17151355 43 1 pair
17161355 44 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 17191355 Supplier ABEBA™ Supplier No. 37328/47
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
Anthracite dynamic occupational clogs with non-foldable heel strap, replaceable active comfort insole, slip resistant TPU outsole
  • Breathable, lightweight, comfortable occupational clogs
  • Comply to EN ISO 20347: 2012, OB SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • Water repellent anthracite knitted textile
  • Non foldable heel strap, adjustable
  • Removable active comfort insole
Specifications
Color Anthracite
Material Knitted Fabric
Size 47
Certifications/Compliance (Europe) ESD dissipative to EN 61340 Climate class 1, Comply to EN ISO 20347: 2012 OB SRC
Gender Unisex
Ankle Style Clogs
Waterproof Water Resistant
Product Type OB SRC ESD